siRNA Details
Gene Symbol: Zfp961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164581.1 | NP_001158053.1 | 4 | 409 | ||
GenBank mRNA | AK142417.1 | - | 1 | 1267 | ||
AK158413.1 | - | 4 | 407 | |||
BC049349.1 | - | 3 | 305 |
