siRNA Details
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115130.1 | NP_001108602.1 | 2 | 698 | ||
NM_172765.3 | NP_766353.2 | 2 | 698 | |||
GenBank mRNA | AK036895.1 | - | 2 | 698 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbtb44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115130.1 | NP_001108602.1 | 2 | 698 | ||
NM_172765.3 | NP_766353.2 | 2 | 698 | |||
GenBank mRNA | AK036895.1 | - | 2 | 698 |