siRNA Details
Gene Symbol: Olfr149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207138.1 | NP_997021.1 | 1 | 921 | ||
GenBank mRNA | BC141260.1 | - | 1 | 976 | 9055872 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207138.1 | NP_997021.1 | 1 | 921 | ||
GenBank mRNA | BC141260.1 | - | 1 | 976 | 9055872 |