siRNA Details
Gene Symbol: Nbeal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183276.2 | NP_899099.2 | 49 | 7786 | ||
GenBank mRNA | AK129158.1 | - | 24 | 4054 | ||
BC157956.1 | - | 49 | 7767 | 9007284 | ||
BC172118.1 | - | 48 | 7686 | 9054404 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC052551.1 | - | 329 | Chr. - - [-] on Build |
