siRNA Details
Gene Symbol: Xcr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011798.4 | NP_035928.2 | 2 | 709 | ||
GenBank mRNA | AK040878.1 | - | 2 | 710 | ||
AK089908.1 | - | 2 | 710 | |||
BC141368.1 | - | 1 | 616 | 9055981 | ||
BC141369.1 | - | 1 | 616 | 9055982 |
