siRNA Details
Gene Symbol: Zfp458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001152.2 | NP_001001152.1 | 4 | 1660 | ||
GenBank mRNA | AK133308.1 | - | 1578 | |||
BC062958.1 | - | 4 | 1660 | 30548236 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001152.2 | NP_001001152.1 | 4 | 1660 | ||
GenBank mRNA | AK133308.1 | - | 1578 | |||
BC062958.1 | - | 4 | 1660 | 30548236 |