siRNA Details
Gene Symbol: Kctd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177715.4 | NP_808383.3 | 1 | 1132 | ||
GenBank mRNA | AK035535.1 | - | 1 | 1134 | ||
AK172581.1 | - | 1 | 1133 | |||
AK173260.1 | - | 1 | 637 | |||
BC116647.1 | - | 2 | 519 | |||
BC116648.1 | - | 2 | 519 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006252420.
Gene Ontology Categories:
