siRNA Details
Gene Symbol: Kctd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177715.4 | NP_808383.3 | 1 | 467 | ||
GenBank mRNA | AK035535.1 | - | 1 | 468 | ||
AK087895.1 | - | 1 | 470 | |||
AK172581.1 | - | 1 | 468 | |||
CF538046.1 | - | 1 | 253 |
