siRNA Details
Gene Symbol: Card6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163138.1 | NP_001156610.1 | 4 | 3067 | ||
GenBank mRNA | AK052796.1 | - | 3 | 2957 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Card6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163138.1 | NP_001156610.1 | 4 | 3067 | ||
GenBank mRNA | AK052796.1 | - | 3 | 2957 |