siRNA Details
Gene Symbol: AU021092 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033220.3 | NP_001028392.2 | 8 | 1025 | ||
GenBank mRNA | AK140126.1 | - | 8 | 1006 | ||
BC150828.1 | - | 8 | 1005 | 9087739 | ||
BC171979.1 | - | 8 | 1005 | 9054265 | ||
BX635104.1 | - | 1 | 347 |
