siRNA Details
Gene Symbol: Zfp407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033341.2 | NP_001028513.2 | 2 | 1686 | ||
GenBank mRNA | AK032536.1 | - | 2 | 1687 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033341.2 | NP_001028513.2 | 2 | 1686 | ||
GenBank mRNA | AK032536.1 | - | 2 | 1687 |