siRNA Details
Gene Symbol: Spry4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011898.2 | NP_036028.2 | 2 | 771 | ||
GenBank mRNA | AB019280.1 | - | 2 | 840 | ||
AF176906.1 | - | 2 | 924 | |||
AK037180.1 | - | 2 | 846 | |||
AK048122.1 | - | 2 | 849 | |||
BC050944.1 | - | 2 | 771 | 6821944 | ||
BC057005.1 | - | 2 | 771 | 6821944 |
