siRNA Details
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112705.2 | NP_001106176.1 | 17 | 2042 | ||
NM_001294329.1 | NP_001281258.1 | 15 | 1902 | |||
NM_001294330.1 | NP_001281259.1 | 15 | 1850 | |||
NM_001294331.1 | NP_001281260.1 | 17 | 2055 | |||
NM_001294334.1 | NP_001281263.1 | 17 | 2042 | |||
GenBank mRNA | AF045252.1 | - | 17 | 2061 | ||
AF045253.1 | - | 16 | 1489 | |||
AF045254.1 | - | 12 | 1203 | |||
AK014829.1 | - | 17 | 1680 | |||
AK089925.1 | - | 15 | 1430 | |||
AK165667.1 | - | 17 | 2045 | |||
AK202282.1 | - | 35 | ||||
AK211763.1 | - | 1 | 36 | |||
AX775743.1 | - | 11 | 1067 | |||
BC066198.1 | - | 15 | 1395 | |||
U56909.1 | - | 8 | 740 |
