siRNA Details
Gene Symbol: Impad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177730.3 | NP_808398.1 | 1 | 461 | ||
GenBank mRNA | AK045511.1 | - | 1 | 511 | ||
BC048776.1 | - | 1 | 461 | 6439769 | ||
BC138209.1 | - | 1 | 318 | 8861228 | ||
BC145952.1 | - | 1 | 318 | 40131255 |
Gene Symbol: LOC100036540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052165.1 | - | 1 | 226 |
