siRNA Details
Gene Symbol: 1700024P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162980.1 | NP_001156452.1 | 2 | 940 | ||
NM_001285952.1 | NP_001272881.1 | 2 | 940 | |||
NM_001285953.1 | NP_001272882.1 | 2 | 873 | |||
GenBank mRNA | AK028077.1 | - | 2 | 880 | ||
AK133762.1 | - | 2 | 811 | |||
BC137723.1 | - | 2 | 940 | 8860742 | ||
BC144857.1 | - | 2 | 940 | 9053405 | ||
BC144858.1 | - | 2 | 941 | 9053406 |
