siRNA Details
Gene Symbol: E2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087452.1 | - | 4 | 1063 | ||
AK157322.1 | - | 1 | 649 | |||
AK157676.1 | - | 2 | 213 | |||
BC062101.1 | - | 4 | 770 | 6848237 | ||
BY231313.1 | - | 2 | 236 |
