siRNA Details
Gene Symbol: E2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087452.1 | - | 2 | 724 | ||
BC062101.1 | - | 2 | 431 | 6848237 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK087452.1 | - | 2 | 724 | ||
BC062101.1 | - | 2 | 431 | 6848237 |