siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 2 | 1097 | ||
GenBank mRNA | AK090326.1 | - | 2 | 1081 | ||
BC070441.1 | - | 2 | 1029 | 30535318 | ||
BC072569.1 | - | 2 | 1062 | 5719275 |
