siRNA Details
Gene Symbol: Ptchd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083342.1 | NP_001076811.1 | 3, 4 | 1417 | ||
GenBank mRNA | AB235901.1 | - | 2, 3 | 1171 | ||
AK090326.1 | - | 3, 4 | 1401 | |||
AK173152.1 | - | 2, 3 | 860 | |||
BC070441.1 | - | 3, 4 | 1349 | 30535318 | ||
BC072569.1 | - | 3, 4 | 1382 | 5719275 |
