siRNA Details
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172891.2 | NP_766479.1 | 2 | 971 | ||
GenBank mRNA | AK033606.1 | - | 2 | 971 | ||
AK035247.1 | - | 3 | 399 | |||
AY563053.1 | - | 2 | 126 | |||
BC150724.1 | - | 2 | 179 | 9087635 |
