siRNA Details
Gene Symbol: Phlpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122594.2 | NP_001116066.2 | 15 | 2376 | ||
GenBank mRNA | AK048260.1 | - | 15 | 2333 | ||
BC117961.1 | - | 15 | 2376 | 40098281 | ||
BC117962.1 | - | 15 | 2368 | 40098282 |
