siRNA Details
Gene Symbol: Phlpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122594.2 | NP_001116066.2 | 19 | 3811 | ||
GenBank mRNA | AK048260.1 | - | 19 | 3768 | ||
BC117961.1 | - | 19 | 3811 | 40098281 | ||
BC117962.1 | - | 19 | 3803 | 40098282 |
