siRNA Details
Gene Symbol: Atr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019864.1 | NP_063917.1 | 4 | 1248 | ||
GenBank mRNA | AK135538.1 | - | 4 | 1146 | ||
AK139834.1 | - | 4 | 1226 | |||
AK142244.1 | - | 4 | 1208 | |||
AK157754.1 | - | 4 | 1196 | |||
AK157757.1 | - | 4 | 1248 | |||
AK162661.1 | - | 4 | 1207 | |||
AK170039.1 | - | 4 | 1177 | |||
AK171072.1 | - | 4 | 1180 |
