siRNA Details
Gene Symbol: Atr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019864.1 | NP_063917.1 | 9 | 2105 | ||
GenBank mRNA | AK139834.1 | - | 9 | 2083 | ||
AK142244.1 | - | 9 | 2066 | |||
AK157754.1 | - | 9 | 2053 | |||
AK171072.1 | - | 9 | 2037 |
