siRNA Details
Gene Symbol: Atr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019864.1 | NP_063917.1 | 45 | 7755 | ||
GenBank mRNA | AF236887.1 | - | 4 | 526 | ||
AK132223.1 | - | 19 | 2880 | |||
AK220176.1 | - | 17 | 2561 |
