siRNA Details
Gene Symbol: Olfr218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001809.2 | NP_001001809.2 | 1 | 248 | ||
GenBank mRNA | BC104284.1 | - | 1 | 247 | ||
BC104291.1 | - | 1 | 248 | 40042773 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001809.2 | NP_001001809.2 | 1 | 248 | ||
GenBank mRNA | BC104284.1 | - | 1 | 247 | ||
BC104291.1 | - | 1 | 248 | 40042773 |