siRNA Details
Gene Symbol: Nav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081035.1 | NP_001074504.1 | 11 | 2453 | ||
GenBank mRNA | AK122404.1 | - | 1 | 284 | ||
BC046396.1 | - | 5 | 966 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081035.1 | NP_001074504.1 | 11 | 2453 | ||
GenBank mRNA | AK122404.1 | - | 1 | 284 | ||
BC046396.1 | - | 5 | 966 |