siRNA Details
Gene Symbol: Gm1564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127576.2 | NP_001121048.1 | 3, 4 | 559 | ||
GenBank mRNA | AK131737.1 | - | 357 | |||
AK163979.1 | - | 3, 4 | 561 | |||
BB610289.1 | - | 357 |
