siRNA Details
Gene Symbol: Lsamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175548.3 | NP_780757.1 | 1 | 51 | ||
GenBank mRNA | AK030681.1 | - | 1 | 356 | ||
AK044845.1 | - | 1 | 51 | |||
AK140043.1 | - | 1 | 188 | |||
AK140663.1 | - | 1 | 50 | |||
AK158702.1 | - | 1 | 535 | |||
BC059067.1 | - | 1 | 567 |
