siRNA Details
Gene Symbol: Cln8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012000.3 | NP_036130.1 | 2, 3 | 1253 | ||
GenBank mRNA | AF125307.1 | - | 2, 3 | 743 | ||
AF125308.1 | - | 2, 3 | 743 | |||
AK049145.1 | - | 2, 3 | 926 | |||
AK080049.1 | - | 2, 3 | 761 | |||
AK088300.1 | - | 2, 3 | 907 | |||
AK089692.1 | - | 2, 3 | 902 | |||
AK128890.1 | - | 2, 3 | 924 | |||
AK164268.1 | - | 2, 3 | 926 | |||
AK166905.1 | - | 2, 3 | 1678 | |||
AK169698.1 | - | 2, 3 | 907 | |||
AK171446.1 | - | 2, 3 | 935 | |||
BC021625.1 | - | 2, 3 | 759 | 4188527 |
