siRNA Details
Gene Symbol: Map4k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252200.1 | NP_001239129.1 | 17, 18 | 2307 | ||
NM_008696.2 | NP_032722.2 | 16, 17 | 2145 | |||
GenBank mRNA | AK129191.1 | - | 16, 17 | 1976 | ||
BC145390.1 | - | 17, 18 | 2041 | 9053948 | ||
U88984.1 | - | 16, 17 | 1825 |
