siRNA Details
Gene Symbol: Map4k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252200.1 | NP_001239129.1 | 6 | 812 | ||
NM_001252201.1 | NP_001239130.1 | 6 | 812 | |||
NM_001252202.1 | NP_001239131.1 | 6 | 812 | |||
NM_008696.2 | NP_032722.2 | 6 | 812 | |||
GenBank mRNA | AK129191.1 | - | 6 | 549 | ||
BC056650.1 | - | 6 | 555 | |||
BC141109.1 | - | 6 | 546 | 9055721 | ||
BC145390.1 | - | 6 | 546 | 9053948 | ||
BC145391.1 | - | 6 | 545 | 9053949 | ||
CF531764.1 | - | 4 | 370 | |||
U88984.1 | - | 6 | 492 |
