siRNA Details
Gene Symbol: Zfp512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172993.3 | NP_766581.2 | 11 | 1321 | ||
GenBank mRNA | AK084205.1 | - | 11 | 1268 | ||
AK146353.1 | - | 11 | 1285 | |||
AK149921.1 | - | 11 | 1268 | |||
AK154918.1 | - | 11 | 1268 | |||
AK173267.1 | - | 11 | 1296 | |||
BC021520.1 | - | 9 | 1115 | |||
BC066136.1 | - | 10 | 1074 | |||
BC089160.1 | - | 11 | 1243 | 5390706 |
