siRNA Details
Gene Symbol: AU019823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134902.1 | NP_001128374.1 | 5 | 557 | ||
NM_212449.2 | NP_997614.2 | 4 | 530 | |||
GenBank mRNA | AK133777.1 | - | 4 | 904 | ||
BC067205.1 | - | 4 | 312 | 6335744 | ||
BY740784.1 | - | 5 | 374 |
