siRNA Details
Gene Symbol: Tceb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013736.4 | NP_038764.2 | 4 | 826 | ||
GenBank mRNA | AB025015.1 | - | 4 | 810 | ||
AK036592.1 | - | 4 | 827 | |||
AK145994.1 | - | 4 | 827 | |||
AK147088.1 | - | 4 | 821 | |||
BC049885.1 | - | 4 | 822 | 5387774 |
