siRNA Details
Gene Symbol: Rilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029938.2 | NP_001025109.1 | 4 | 609 | ||
GenBank mRNA | AK139016.1 | - | 4 | 601 | ||
AK160517.1 | - | 2 | 402 | |||
BC051502.1 | - | 1 | 68 | |||
BC116750.1 | - | 4 | 593 | 40126069 | ||
BC119166.1 | - | 4 | 593 | 8733915 |
