siRNA Details
Gene Symbol: Tor3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023141.2 | NP_075630.2 | 3 | 418 | ||
GenBank mRNA | AJ299404.1 | - | 3 | 386 | ||
AK007814.1 | - | 3 | 407 | |||
AK009693.1 | - | 3 | 436 | |||
AK017908.1 | - | 3 | 363 | |||
AK151048.1 | - | 3 | 404 | |||
AK151390.1 | - | 3 | 385 | |||
AK152243.1 | - | 3 | 383 | |||
AK152358.1 | - | 3 | 419 | |||
AK152484.1 | - | 3 | 419 | |||
AK153391.1 | - | 3 | 404 | |||
AK170296.1 | - | 3 | 413 | |||
AK172355.1 | - | 3 | 393 | |||
BC025525.1 | - | 3 | 403 | |||
BC052851.1 | - | 3 | 707 | |||
BC071216.1 | - | 3 | 399 | 30439199 |
