siRNA Details
Gene Symbol: Pianp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145926.1 | NP_001139398.1 | 3 | 480 | ||
NM_001145927.1 | NP_001139399.1 | 3 | 464 | |||
NM_175696.4 | NP_783627.2 | 3 | 635 | |||
GenBank mRNA | AK082984.1 | - | 3 | 637 | ||
AK148352.1 | - | 3 | 481 | |||
AK187032.1 | - | 1 | 150 | |||
AK202216.1 | - | 1 | 150 | |||
BC065170.1 | - | 3 | 464 | 6413408 |
