siRNA Details
Gene Symbol: Mpzl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093749.2 | NP_001087218.1 | 1 | 122 | ||
GenBank mRNA | AK041037.1 | - | 1 | 116 | ||
AK049878.1 | - | 1 | 114 | |||
BC132150.1 | - | 1 | 105 | |||
EF102773.1 | - | 1 | 86 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK040091.1 | - | 39 | Chr. - - [-] on Build |

More Information
Other Species Targeted

Gene Ontology Categories:
