siRNA Details
Gene Symbol: Zfp865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033383.2 | NP_001028555.1 | 2 | 3047 | ||
GenBank mRNA | AK031793.1 | - | 1 | 2034 | ||
AK032361.1 | - | 1 | 2432 | |||
AK154737.1 | - | 2 | 3048 |
