siRNA Details
Gene Symbol: Tmem72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178768.4 | NP_848883.2 | 2, 3 | 310 | ||
GenBank mRNA | AK033856.1 | - | 2, 3 | 446 | ||
AK049048.1 | - | 2, 3 | 465 | |||
AK049938.1 | - | 2, 3 | 423 | |||
AK085603.1 | - | 2, 3 | 311 | |||
AK141578.1 | - | 2, 3 | 467 | |||
BC093486.1 | - | 2, 3 | 453 | 30697044 | ||
BC116440.1 | - | 2, 3 | 294 | 40104615 | ||
BC116441.1 | - | 2, 3 | 294 | 40104616 |
