siRNA Details
Gene Symbol: Dock6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177030.3 | NP_796004.2 | 14, 15 | 1685 | ||
GenBank mRNA | AK129348.1 | - | 12, 13 | 1746 | ||
AK145109.1 | - | 14, 15 | 1669 | |||
BU705495.1 | - | 1, 2 | 133 |
