siRNA Details
Gene Symbol: Dock6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177030.3 | NP_796004.2 | 3 | 293 | ||
GenBank mRNA | AK049320.1 | - | 3 | 275 | ||
AK080190.1 | - | 3 | 275 | |||
AK129348.1 | - | 1 | 363 | |||
AK145109.1 | - | 3 | 277 | |||
CJ099751.1 | - | 3 | 293 |
