siRNA Details
Gene Symbol: Ccdc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_180958.3 | NP_851289.2 | 10 | 995 | ||
GenBank mRNA | AB775896.1 | - | 8 | 776 | ||
AK029754.1 | - | 10 | 996 | |||
AK030157.1 | - | 9 | 1309 | |||
BC113771.1 | - | 9 | 1319 | |||
BC129814.1 | - | 9 | 1320 |
