siRNA Details
Gene Symbol: Itpkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081175.1 | NP_001074644.1 | 4 | 2546 | ||
GenBank mRNA | AK039369.1 | - | 3 | 461 | ||
BC151160.1 | - | 4 | 2412 | 9088049 | ||
BC157986.1 | - | 4 | 2411 | 9088142 | ||
BC158000.1 | - | 4 | 2411 | 9088148 |
