siRNA Details
Gene Symbol: Gfm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146043.2 | NP_001139515.1 | 6 | 516 | ||
NM_001271463.1 | NP_001258392.1 | 6 | 516 | |||
NM_001271464.1 | NP_001258393.1 | 6 | 516 | |||
NM_001271465.1 | NP_001258394.1 | 6 | 543 | |||
NM_177266.5 | NP_796240.3 | 6 | 549 | |||
GenBank mRNA | AK028450.1 | - | 6 | 502 | ||
AK035819.1 | - | 6 | 455 | |||
AK044371.1 | - | 6 | 517 | |||
AK083441.1 | - | 6 | 481 | |||
AK161797.1 | - | 6 | 452 | |||
AK165871.1 | - | 6 | 510 | |||
AK219489.1 | - | 5 | 362 | |||
BC027341.1 | - | 6 | 500 | 3984129 | ||
BC138233.1 | - | 6 | 447 | 8861252 | ||
BC145208.1 | - | 6 | 447 | 9053759 | ||
BY742160.1 | - | 6 | 517 |
