siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 1966 | ||
GenBank mRNA | AK030106.1 | - | 6 | 1968 | ||
AK122469.1 | - | 1 | 139 | |||
BC053480.1 | - | 1 | 657 | |||
BC086807.1 | - | 1 | 585 |
