siRNA Details
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 641 | ||
GenBank mRNA | AK030106.1 | - | 6 | 642 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C530008M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163793.1 | NP_001157265.1 | 6 | 641 | ||
GenBank mRNA | AK030106.1 | - | 6 | 642 |