siRNA Details
Gene Symbol: Atp11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001798.2 | NP_001001798.1 | 13 | 1459 | ||
NM_001037863.1 | NP_001032952.1 | 13 | 1459 | |||
GenBank mRNA | AK043065.1 | - | 13 | 1453 | ||
AK143860.1 | - | 12 | 1337 | |||
BC106087.1 | - | 13 | 1438 | 30843359 |

More Information
Other Species Targeted

Gene Ontology Categories:
