siRNA Details
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177815.3 | NP_808483.2 | 11 | 1229 | ||
GenBank mRNA | BC082539.1 | - | 11 | 1204 | ||
BC141275.1 | - | 11 | 1223 | 9055887 | ||
BC145606.1 | - | 11 | 1136 | 9054172 |
